Learn More
Zeiss™ Axio Vert.A1 Inverted Microscope for Phase Contrast and DIC
Product Code. 17312830
Click to view available options
Includes:
Phase Contrast and DIC
Product Code. 17312830
Brand: Zeiss™ 4912370021000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A compact inverted microscope ideal for routine research. This system is configured for transmitted light brightfield, phase contrast and DIC with a binocular phototube. The Axio Vert.A1 offers flexibility and upgradability with additional contrast techniques and a range of accessories available.
  • The Axio Vert.A1 is an inverted microscope ideal for advanced routine research and is the only system in its class with such a large range of features but still compact enough to sit directly beside your incubator. The system offers the ultimate flexibility and all standard contrast techniques including brightfield, phase contrast, PlasDIC, iHMC, DIC and epi-fluorescence are available
  • The system has been designed with ergonomics and ease of use in mind. Whether you are sitting or standing, simply use intermediate pieces to work comfortably in an upright position. The ergo tube lets you tilt the eyepieces as much as you need. You can always view your samples easily. Thanks to ergonomic design of all operating parts, your routine tasks and workflows will be easier
  • The Axio Vert.A1 offers either Halaogen or LED illumination for transmitted light. There are then two options for epi-fluorescence - either the Axio Vert.A1 FL-LED with inbuilt LEDs or the Axio Vert.A1 Fl with the ability to use an external light source for fluorescence to give more flexibility.
  • The accessories available with the Axio Vert.A1 are vast and these include multiple binocular tubes, documentation ports and attachments for infinity space. There are a range of options for stages with inserts available for a range of samples.
Specifications
Includes Phase Contrast and DIC