Learn More
Zeiss™ Axio Vert.A1 Inverted Microscope for Phase Contrast
Product Code. p-7235929
Change view
Click to view available options
Condenser Type:
LD Condenser 0.3
LD Condenser 0.4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Condenser Type
17372820 LD Condenser 0.3
17382820 LD Condenser 0.4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17372820 Supplier Zeiss™ Supplier No. 4912370001000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A compact inverted microscope ideal for routine research. This system is configured for transmitted light brightfield and phase contrast.
  • The Axio Vert.A1 is an inverted microscope ideal for advanced routine research and is the only system in its class with such a large range of features but still compact enough to sit directly beside your incubator. The system offers the ultimate flexibility and all standard contrast techniques including brightfield, phase contrast, PlasDIC, iHMC, DIC and epi-fluorescence are available.
  • The system has been designed with ergonomics and ease of use in mind. Whether you are sitting or standing, simply use intermediate pieces to work comfortably in an upright position. The ergo tube lets you tilt the eyepieces as much as you need. You can always view your samples easily. Thanks to ergonomic design of all operating parts, your routine tasks and workflows will be easier.
  • The Axio Vert.A1 offers either Halaogen or LED illumination for transmitted light. There are then two options for epi-fluorescence - either the Axio Vert.A1 FL-LED with inbuilt LEDs or the Axio Vert.A1 Fl with the ability to use an external light source for fluorescence to give more flexibility.
  • The accessories available with the Axio Vert.A1 are vast and these include multiple binocular tubes, documentation ports and attachments for infinity space. There are a range of options for stages with inserts available for a range of samples.
Specifications
Condenser Type LD Condenser 0.3
For Use With (Application) Phase Contrast