missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AXIN2 Recombinant Rabbit Monoclonal Antibody (23GB1545), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | AXIN2 |
|---|---|
| Clone | 23GB1545 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089467
![]() |
Invitrogen™
MA552730 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088796
|
Invitrogen™
MA552731 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AXIN2 Recombinant Monoclonal antibody specifically detects AXIN2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| AXIN2 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O70240, O88566, Q9Y2T1 | |
| 12006, 29134, 8313 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1545 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Axi1; axil; axin 2; Axin like protein; axin2; axin-2; Axin-like protein; Axis inhibition protein 2; conductin; DKFZp781B0869; MGC10366; MGC126582; ODCRCS | |
| A synthesized peptide derived from human Axin2 (58-93AA). | |
| Primary | |
| -20°C | |
| AXIN2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title