missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AXIN2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521093
This item is not returnable.
View return policy
Description
A suggested positive control is mouse lung tissue lysate. PA5-21093 can be used with blocking peptide PEP-1207.
The Axin-related protein, Axin2, presumably plays an important role in the regulation of the stability of beta-catenin in the Wnt signaling pathway, like its rodent homologs, mouse conductin/rat axil. In mouse, conductin organizes a multiprotein complex of APC (adenomatous polyposis of the colon), beta-catenin, glycogen synthase kinase 3-beta, and conductin, which leads to the degradation of beta-catenin. Apparently, the deregulation of beta-catenin is an important event in the genesis of a number of malignancies. The AXIN2 gene has been mapped to 17q23-q24, a region that shows frequent loss of heterozygosity in breast cancer, neuroblastoma, and other tumors. Mutations in this gene have been associated with colorectal cancer with defective mismatch repair.
Specifications
| AXIN2 | |
| Polyclonal | |
| Unconjugated | |
| AXIN2 | |
| Axi1; axil; axin 2; Axin like protein; axin2; axin-2; Axin-like protein; Axis inhibition protein 2; conductin; DKFZp781B0869; MGC10366; MGC126582; ODCRCS | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12006, 29134, 8313 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| O70240, O88566, Q9Y2T1 | |
| AXIN2 | |
| A 20 amino acid peptide near the carboxy terminus of human AXIN2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction