missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AXIN1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16541-1-AP-20UL
This item is not returnable.
View return policy
Description
Proteintech's AXIN1 antibody 16541-1-AP is a rabbit polyclonal antibody raised against the N-terminus of human AXIN1.
AXIN1 is a cytoplasmic protein which contains a regulation of G-protein signaling (RGS) domain and a disheveled and axin (DIX) domain and is thought to function as a negative regulator of the WNT signaling pathway that regulates embryonic axis formation. AXIN1 interacts with adenomatosis polyposis coli (APC), beta-catenin, glycogen synthase kinase 3 beta, forming a tetrameric complex resulting in the regulation of the stabilization of beta-catenin. Mutations in the AXIN1 gene have been associated various carcinomas, indicating that it also functions as a tumor suppressor.Specifications
| AXIN1 | |
| Polyclonal | |
| Unconjugated | |
| AXIN1 | |
| AXIN, axin 1, AXIN1, Axis inhibition protein 1, hAxin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8312 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O15169 | |
| AXIN1 | |
| AXIN1 Fusion Protein Ag9858 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction