missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axin-2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76526-0.025mg
This item is not returnable.
View return policy
Description
Axin-2 Polyclonal specifically detects Axin-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Axin-2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Axil, axin 2, axin-2, Axin-like protein, Axis inhibition protein 2, Conductin, DKFZp781B0869, MGC10366, MGC126582 | |
| Rabbit | |
| 85, 93 kDa | |
| 0.025 mg | |
| Signal Transduction, Wnt Signaling Pathway | |
| 8313 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL, Immunofluorescence 20 μg/mL | |
| AAF22799 | |
| AXIN2 | |
| Antibody was raised against a 20 amino acid synthetic peptide near the carboxy terminus of human AXIN2. The immunogen is located within amino acids 780 - 830 of AXIN2. Amino Acid Squence: KEQLSKKGNYRYYFKKASDE | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Axin-2 antibody is not predicted to cross-react with AXIN1. At least four isoforms of AXIN2 are known to exist; this antibody will recognize all four. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction