missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axin-2 Antibody (3G6), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | Axin-2 |
|---|---|
| Clone | 3G6 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30032044
|
Novus Biologicals
NBP3-26485-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30033733
|
Novus Biologicals
NBP3-26485-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Axin-2 Monoclonal antibody specifically detects Axin-2 in Human samples. It is validated for ELISASpecifications
| Axin-2 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Axil, axin 2, axin-2, Axin-like protein, Axis inhibition protein 2, Conductin, DKFZp781B0869, MGC10366, MGC126582 | |
| A synthesized peptide derived from Human Axin-2 [UniProt Q9Y2T1] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 3G6 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction, Wnt Signaling Pathway | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 8313 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title