missing translation for 'onlineSavingsMsg'
Learn More

Axin-1 Antibody, Novus Biologicals™

Product Code. 18394265 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394265 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18394265 Supplier Novus Biologicals Supplier No. NBP318242

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Axin-1 Polyclonal antibody specifically detects Axin-1 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Axin-1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS pH 7.4, 50% glycerol
Gene Alias axin 1, axin-1, AXINaxis inhibitor 1, Axis inhibition protein 1, fused, mouse, homolog of, hAxin, MGC52315
Host Species Rabbit
Immunogen Synthetic peptide of Human Axin-1 (200-300 aa), conjugated to Keyhole Limpet Haemocyanin (KLH).
Purification Method Peptide affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8312
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.