missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axin-1 Antibody (4E9F1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£176.00 - £435.00
Tekniske data
| Antigen | Axin-1 |
|---|---|
| Clone | 4E9F1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:250 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|
18273035
|
Novus Biologicals
NBP2-61695 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18615688
|
Novus Biologicals
NBP2-61695-0.025ml |
0.025 mL |
£176.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
Axin-1 Monoclonal specifically detects Axin-1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Tekniske data
| Axin-1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| axin 1, axin-1, AXINaxis inhibitor 1, Axis inhibition protein 1, fused, mouse, homolog of, hAxin, MGC52315 | |
| AXIN1 | |
| IgG2b | |
| Protein G purified |
| 4E9F1 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:50-1:250 | |
| Monoclonal | |
| Purified | |
| Apoptosis, Wnt Signaling Pathway | |
| 8312 | |
| Purified recombinant fragment of human AXIN1 (AA: 546-752) expressed in E. Coli. | |
| Primary | |
| 95.6 kDa |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel