missing translation for 'onlineSavingsMsg'
Learn More

AX20017, MedChemExpress

Product Code. 16486278
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486278 10 mg 10mg
16453456 25 mg 25mg
16448867 5 mg 5mg
16432407 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16486278 Supplier Medchem Express Supplier No. HY14987

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AX20017 is a small-molecule protein kinase G (PknG) inhibitor with an IC50 of 0.39 μM.

AX20017 is a small-molecule protein kinase G (PknG) inhibitor with an IC50 of 0.39 μM.

Specifications

Chemical Name or Material AX20017
Physical Form Solid
CAS 329221-38-7
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C13H16N2O2S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 130 mg/mL (491.79 mM; Need ultrasonic)
SMILES O=C(N)C1=C(NC(C2CC2)=O)SC3=C1CCCC3
Molecular Weight (g/mol) 264.34
Formula Weight 264.34
Percent Purity 99.88%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.