Learn More
Invitrogen™ AWAT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113790
Description
Immunogen sequence: GEPLPMPKIE NPSQEIVAKY HTLYIDALRK LFDQHKTKFG ISETQELEII For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Dog: 86%; Guinea Pig: 86%; Horse: 79%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%.
This gene encodes an enzyme belonging to the diacylglycerol acyltransferase family. This enzyme produces wax esters by the esterification of long chain (or wax) alcohols with acyl-CoA-derived fatty acids. It functions in lipid metabolism in the skin, mostly in undifferentiated peripheral sebocytes. This enzyme may also have acyl-CoA:retinol acyltransferase activities, where it catalyzes the synthesis of diacylglycerols and retinyl esters.
Specifications
| AWAT2 | |
| Polyclonal | |
| Unconjugated | |
| AWAT2 | |
| 11-cis-RE-synthase; 11-cis-specific retinyl-ester synthase; 9430062J17Rik; acyl-CoA retinol O-fatty-acyltransferase; Acyl-CoA wax alcohol acyltransferase 2; ARAT; Awat2; DC4; DGAT2L4; diacylglycerol O-acyltransferase 2-like 4; diacylglycerol O-acyltransferase 2-like protein 4; Diacylglycerol O-acyltransferase candidate 4; hDC4; hWS; Long-chain-alcohol O-fatty-acyltransferase 2; MFAT; Multifunctional O-acyltransferase; mWS; retinol O-fatty-acyltransferase; RGD1565111; Wax synthase; wax synthase 2; WS | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 158835 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6E213 | |
| AWAT2 | |
| Synthetic peptide directed towards the C terminal region of human DGAT2L4 (aa 284-333). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.