missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AWAT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550948
This item is not returnable.
View return policy
Description
AWAT1 Polyclonal Antibody detects endogenous levels of AWAT1 protein.
The protein encoded by this gene belongs to the diacylglycerol acyltransferase family. It esterifies long chain (wax) alcohols with acyl-CoA-derived fatty acids to produce wax esters. Wax esters are enriched in sebum, suggesting that this enzyme plays a central role in lipid metabolism in skin. Consistent with this observation, this protein is predominantly expressed in the sebaceous gland of the skin. [provided by RefSeq, Sep 2009].
Specifications
| AWAT1 | |
| Polyclonal | |
| Unconjugated | |
| AWAT1 | |
| Acyl-CoA wax alcohol acyltransferase 1; AWAT1; DGA2; Dgat2l3; diacylglycerol acyltransferase 2; diacyl-glycerol acyltransferase 2; diacylglycerol O-acyltransferase 2-like 3; diacylglycerol O-acyltransferase 2-like protein 3; Long-chain-alcohol O-fatty-acyltransferase 1; wax synthase 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 158833 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q58HT5 | |
| AWAT1 | |
| Synthesized peptide derived from the C-terminal region of human DGAT2L3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction