missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AWAT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59865
This item is not returnable.
View return policy
Description
AWAT1 Polyclonal specifically detects AWAT1 in Human samples. It is validated for Western Blot.
Specifications
| AWAT1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| acyl-CoA wax alcohol acyltransferase 1, DGA2, DGAT2L3, diacyl-glycerol acyltransferase 2, Diacylglycerol acyltransferase 2, diacylglycerol O-acyltransferase 2-like 3, Diacylglycerol O-acyltransferase 2-like protein 3, EC 2.3.1.75, Long-chain-alcohol O-fatty-acyltransferase 1 | |
| Rabbit | |
| 38 kDa | |
| 100 μL | |
| Primary | |
| Expected to cross react based on sequence identity: Porcine 85%, Equine 86%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q58HT5 | |
| AWAT1 | |
| Synthetic peptides corresponding to AWAT1 (acyl-CoA wax alcohol acyltransferase 1) Antibody(against the N terminal of AWAT1. Peptide sequence NWCVWTHIRDYFPITILKTKDLSPEHNYLMGVHPHGLLTFGAFCNFCTEA. | |
| Affinity purified | |
| RUO | |
| 158833 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction