missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AVPR1B Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA533347

Product Code. 15364461

  • £479.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is not recommended for use in ICC or WB applications. Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Rat, Bovine, Dog, Bat, Elephant, Panda, Horse, Rabbit, Pig, Opossum (100%).

Vasopressin V1B receptor expression has been documented in the pituitary, pituitary tumors, pheochromocytoma, breast cancer, and small-cell lung carcinomas. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more than a thousand members currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This turn-off activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Two blockbuster drugs which have been developed as GPCR-targeted pharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

AVPR1B
Polyclonal
Unconjugated
AVPR1B
Antidiuretic hormone receptor 1b; arginine vasopressin receptor 1B; arginine vasopressin receptor 3; arginine vasopressin receptor V3; arginine vasopressin type 1b receptor; AVP receptor V3; AVPR V1b; AVPR V3; Avpr1b; AVPR3; pituitary vasopressin receptor 3; V1b receptor; V1bR; V3/V1b; V3/V1b pituitary vasopressin receptor; vasopressin V1b receptor; Vasopressin V3 receptor; VIBR; VPR3
Rabbit
Antigen affinity chromatography
RUO
100055260, 100354969, 100517909, 282861, 29462, 488578, 553
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
P47901, P48974
AVPR1B
Synthetic 17 amino acid peptide from N-terminal extracellular domain of human AVPR1B.
50 μg
Primary
Human, Rat, Canine, Rabbit, Bovine, Pig, Horse, Bat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.