Learn More
Invitrogen™ AVPR1A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ 720289
Description
These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. This antibody is predicted to react with Monkey, Cat, Rat and Pig.
Vasopressin receptor 1a (AVPR1A) is a receptor for arginine vasopressin. The activity of this receptor is mediated by G-proteins which activate a phosphatidyl-inositol-calcium second messenger system. AVPR1A has been involved in social behaviors, including affiliation and attachment.
Specifications
| AVPR1A | |
| Polyclonal | |
| Unconjugated | |
| AVPR1A | |
| antidiuretic hormone receptor 1A; arginine vasopressin receptor 1A; arginine vasopressin receptor V1a; AVPR; AVPR V1a; AVPR1; Avpr1a; SCCL vasopressin subtype 1a receptor; V1a; V1a arginine vasopressin receptor; V1a receptor; V1a vasopressin receptor; V1aR; V1-vascular vasopressin receptor AVPR1A; Vascular/hepatic-type arginine vasopressin receptor; vasopressin V1a receptor; Vasopressin V1aR | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 552 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2 | |
| P37288 | |
| AVPR1A | |
| Peptide corresponding to Human AVPR1A (aa 406-418). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.