missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AVP Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA184556

Product Code. 11599170

  • £532.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Rabbit anti arginine vasopressin antibody recognizes arginine vasopressin.

This gene encodes a precursor protein consisting of arginine vasopressin and two associated proteins, neurophysin 2 and a glycopeptide, copeptin. Arginine vasopressin is a posterior pituitary hormone which is synthesized in the supraoptic nucleus and paraventricular nucleus of the hypothalamus. Along with its carrier protein, neurophysin 2, it is packaged into neurosecretory vesicles and transported axonally to the nerve endings in the neurohypophysis where it is either stored or secreted into the bloodstream. The precursor is thought to be activated while it is being transported along the axon to the posterior pituitary. Arginine vasopressin acts as a growth factor by enhancing pH regulation through acid-base transport systems. It has a direct antidiuretic action on the kidney, and also causes vasoconstriction of the peripheral vessels. This hormone can contract smooth muscle during parturition and lactation. It is also involved in cognition, tolerance, adaptation and complex sexual and maternal behaviour, as well as in the regulation of water excretion and cardiovascular functions.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

AVP
Polyclonal
Unconjugated
AVP
ADH; antidiuretic hormone; arginine vasopressin; arginine vasopressin (Diabetes insipidus), same as Di (conflicting physical mapping); arginine vasopressin (neurophysin II, antidiuretic hormone, diabetes insipidus, neurohypophyseal); arginine vasopressin-neurophysin II precursor; Arginine-vasopressin; Arg-vasopressin; ARVP; Avp; AVP-neurophysin II precursor; AVP-NPII; AVRP; Copeptin; DI; neurohypophyseal; Neurophysin 2; Neurophysin-I; Neurophysin-II; prepro-arginine-vasopressin-neurophysin II; prepro-AVP-NP II; prepropressophysin; preprovasopressin-neurophysin; Vas; Vasopressin; vasopressin-neurophysin 2-copeptin; vasopressin-neurophysin II-copeptin; vasopressin-neurophysin prepropeptide; VP; Vsp
Rabbit
100 μL
Primary
Rat
Antibody
IgG
Immunohistochemistry (Frozen)
Conc. Not Determined
Whole Serum, PBS with <0.1% sodium azide
P01186
AVP
Synthetic peptide corresponding to residues CYFQNCPRG of Arginine Vasopression (aa 24-32).
RUO
24221
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.