missing translation for 'onlineSavingsMsg'
Learn More

Avosentan, MedChemExpress

Product Code. 16486358
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486358 10 mg 10mg
16478947 5 mg 5mg
16422477 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16486358 Supplier Medchem Express Supplier No. HY15195

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Avosentan(Ro 67-0565; SPP-301) is a potent, selective endothelin receptor(ETA receptor) antagonist.

Avosentan(Ro 67-0565

Specifications

Chemical Name or Material Avosentan
CAS 290815-26-8
Color White
Purity Grade Notes Research
Molecular Formula C23H21N5O5S
Quantity 10 mg
Synonym Ro 67-0565 SPP-301
Solubility Information DMSO : 6.67 mg/mL (13.91 mM; Need ultrasonic)
SMILES COC1=C(OC(C(NS(C2=NC=C(C=C2)C)(=O)=O)=NC(C3=CC=NC=C3)=N4)=C4OC)C=CC=C1
Molecular Weight (g/mol) 479.51
Formula Weight 479.51
Percent Purity 98.54%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.