missing translation for 'onlineSavingsMsg'
Learn More

Avoralstat, MedChemExpress

Product Code. 16497156
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497156 100 mg 100mg
16490276 2 mg 2mg
16483536 25 mg 25mg
16469697 5 mg 5mg
16467168 10 mg 10mg
16413197 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16497156 Supplier Medchem Express Supplier No. HY16735
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Avoralstat (BCX4161), a potent and orally active plasma kallikrein (PKK) inhibitor, is used for hereditary angioedema research.

Avoralstat (BCX4161), a potent and orally active plasma kallikrein (PKK) inhibitor, is used for hereditary angioedema research.

Specifications

Chemical Name or Material Avoralstat
CAS 918407-35-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C28H27N5O5
Quantity 100 mg
Synonym BCX4161
Solubility Information DMSO : 50 mg/mL (97.36 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(C1=NC(C(NCC2CC2)=O)=CC=C1C3=CC(OC)=C(C=C)C=C3C(NC4=CC=C(C(N)=N)C=C4)=O)O
Molecular Weight (g/mol) 513.54
Formula Weight 513.54
Percent Purity 95.51%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.