missing translation for 'onlineSavingsMsg'
Learn More

Avobenzone, MedChemExpress

Product Code. 16498226
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498226 100 mg 100mg
16472096 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16498226 Supplier Medchem Express Supplier No. HYB0316

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Avobenzone, a dibenzoylmethane compound, is one of the most widely used filters in sunscreens for skin photoprotection in the UVA band. Avobenzone is an endocrine disruptor that directly binds to estrogen receptor β and acts as an estrogen agonist.

Avobenzone, a dibenzoylmethane compound, is one of the most widely used filters in sunscreens for skin photoprotection in the UVA band. Avobenzone is an endocrine disruptor that directly binds to estrogen receptor β and acts as an estrogen agonist.

Specifications

Chemical Name or Material Avobenzone
CAS 70356-09-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H22O3
Quantity 100 mg
Solubility Information DMSO : ≥ 50 mg/mL (161.09 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=CC=C(C(C)(C)C)C=C1)CC(C2=CC=C(OC)C=C2)=O
Molecular Weight (g/mol) 310.39
Formula Weight 310.39
Percent Purity 98.0%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.