missing translation for 'onlineSavingsMsg'
Learn More

AVN-944, MedChemExpress

Product Code. p-200101433
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16488427

Brand: Medchem Express HY13560

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AVN-944 (VX-944) is an orally active, potent, selective, noncompetitive and specific inhibitor of IMPDH (inosine monophosphate dehydrogenase).

AVN-944 is an essential rate-limiting enzyme in de novo guanine nucleotide synthesis. AVN-944 is also an inhibitor of arenavirus RNA synthesis, and blocks arenavirus infection. AVN-944 has broad anti-cancer activities, and can be used for multiple myeloma (MM) and acute myeloid leukemia (AML) research.

Specifications

Chemical Name or Material AVN-944
CAS 297730-17-7
Color White
Purity Grade Notes Research
Molecular Formula C25H27N5O5
Quantity 5 mg
Synonym VX-944
Solubility Information DMSO : 100 mg/mL (209.42 mM; Need ultrasonic)
SMILES O=C(O[C@@H](CC#N)CC)N[C@H](C1=CC=CC(NC(NC2=CC=C(C3=CN=CO3)C(OC)=C2)=O)=C1)C
Molecular Weight (g/mol) 477.51
Formula Weight 477.51
Percent Purity 99.33%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.