missing translation for 'onlineSavingsMsg'
Learn More

AVN-492, MedChemExpress

Product Code. 16464726
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16464726 100 mg 100mg
16440037 50 mg 50mg
16426377 5 mg 5mg
16403648 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16464726 Supplier Medchem Express Supplier No. HY101924
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AVN-492 is a very specific and highly-selective antagonist with picomolar affinity to 5-HT6R (Ki=91 pM).

AVN-492 is a very specific and highly-selective antagonist with picomolar affinity to 5-HT6R (Ki=91 pM).

Specifications

Chemical Name or Material AVN-492
CAS 1220646-23-0
Color White
Purity Grade Notes Research
Molecular Formula C17H21N5O2S
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (278.20 mM)
SMILES O=S(C1=CC=CC=C1)(C2=C3N(C(C)=C(N(C)C)C(C)=N3)N=C2NC)=O
Molecular Weight (g/mol) 359.45
Formula Weight 359.45
Percent Purity 99.27%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.