missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AVIL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107076
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 85%; Horse: 100%; Human: 100%; Mouse: 92%; Pig: 93%; Rabbit: 93%; Rat: 92%.
AVIL is a member of the gelsolin/villin family of actin regulatory proteins. This protein has structural similarity to villin. It binds actin and may play a role in the development of neuronal cells that form ganglia.
Specifications
| AVIL | |
| Polyclonal | |
| Unconjugated | |
| AVIL | |
| actin-binding protein DOC6; Advil; Advillin; Advillin-like protein; Avil; DOC6; p92; peripheral nervous system villin-like protein; pervin; Unknown (protein for MGC:136857); zgc:136857 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10677 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| O75366 | |
| AVIL | |
| Synthetic peptide directed towards the middle region of human AVIL (aa 770-819). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction