missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Avidin HRP

Product Code. 15538306
Click to view available options
Quantity:
500 μL
1 mL
Unit Size:
1mL
500µL
This item is not returnable. View return policy

Product Code. 15538306

Brand: Invitrogen™ 18410094

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Commonly used as a second step for detection of biotinylated antibodies by ELISA and western blot. Avidin binds to biotin with high affinity. [Note: Sodium azide is an inhibitor of horseradish peroxidase enzymatic activity and should not be added to the AV-HRP diluent.]

Avidin binds to biotin with high affinity. Because sodium azide is an inhibitor of horseradish peroxidase enzymatic activity, it should not be added to the AV-HRP diluent.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 500 μL
Target Molecule Biotin
Form Liquid
Product Line eBioscience
Label or Dye HRP (Horseradish Peroxidase)

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.