missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Avidin HRP

Commonly used as a second step for detection of biotinylated antibodies by ELISA and western blot. Avidin binds to biotin with high affinity. [Note: Sodium azide is an inhibitor of horseradish peroxidase enzymatic activity and should not be added to the AV-HRP diluent.]

Brand:  Invitrogen™ 18-4100-51

Code : NEW

Additional Details : Weight : 0.47500kg

 View more versions of this product

Product Code. 15528306

  • £105.00 / 1mL
Estimated Shipment: 03-06-2024
to see stock.
Add to basket

Description

Description

Avidin binds to biotin with high affinity. Because sodium azide is an inhibitor of horseradish peroxidase enzymatic activity, it should not be added to the AV-HRP diluent.

Specifications

Specifications

1 mL
Liquid
HRP (Horseradish Peroxidase)
Biotin
eBioscience™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.