missing translation for 'onlineSavingsMsg'
Learn More

Avibactam sodium, MedChemExpress

Product Code. 16486228
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16486228

Brand: Medchem Express HY14879A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Avibactam sodium (NXL-104) is a covalent and reversible non-β-lactam β-lactamase inhibitor which inhibits β-lactamase TEM-1 and CTX-M-15 with IC50s of 8 nM and 5 nM, respectively.

Avibactam sodium (NXL-104) is a covalent and reversible non-β-lactam β-lactamase inhibitor which inhibits β-lactamase TEM-1 and CTX-M-15 with IC50s of 8 nM and 5 nM, respectively.

Specifications

Chemical Name or Material Avibactam sodium
CAS 1192491-61-4
Color White
Purity Grade Notes Research
Molecular Formula C7H10N3NaO6S
Quantity 10 mg
Synonym NXL-104
Solubility Information H2O : 50 mg/mL (174.08 mM; Need ultrasonic) ∣DMSO : ≥ 30 mg/mL (104.45 mM)
SMILES O=S(ON1[C@]2([H])CC[C@@H](C(N)=O)[N@@](C2)C1=O)(O[Na])=O
Molecular Weight (g/mol) 287.23
Formula Weight 287.23
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.