missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Avian Influenza A H5N1 Neuraminidase Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
£436.00
Specifications
| Antigen | Avian Influenza A H5N1 Neuraminidase |
|---|---|
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 500 SE |
| Form | Purified |
Description
Avian Influenza A H5N1 Neuraminidase Polyclonal antibody specifically detects Avian Influenza A H5N1 Neuraminidase in Virus samples. It is validated for ELISASpecifications
| Avian Influenza A H5N1 Neuraminidase | |
| Polyclonal | |
| Purified | |
| RUO | |
| Virus | |
| IgG | |
| Peptide affinity purified |
| ELISA | |
| mFluor Violet 500 SE | |
| Rabbit | |
| Infections (Virus Bacteria and Parasites) | |
| Antibody was raised against a synthetic peptide corresponding to 15 amino acids near the carboxy terminus of the avian influenza neuraminidase protein. Efforts were made to use relatively conserved regions as the antigen. The immunogen is located within the last 50 amino acids of Avian Influenza Neuraminidase. Amino Acid Squence: IRGRPKENTIWTSGS | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title