missing translation for 'onlineSavingsMsg'
Learn More

Avian Influenza A H5N1 Neuraminidase Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£141.00 - £398.00

Specifications

Antigen Avian Influenza A H5N1 Neuraminidase
Concentration 1 mg/mL
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18690606
View Documents
Novus Biologicals
NBP2-41049-0.025mg
0.025 mg
£141.00
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18195929
View Documents
Novus Biologicals
NBP2-41049
0.1 mg
£398.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Avian Influenza A H5N1 Neuraminidase Polyclonal antibody specifically detects Avian Influenza A H5N1 Neuraminidase in Virus samples. It is validated for ELISA.
Specifications

Specifications

Avian Influenza A H5N1 Neuraminidase
ELISA
Unconjugated
RUO
PBS with 0.02% Sodium Azide
IgG
Affinity Purified
1 mg/mL
Polyclonal
Rabbit
Virus
Antibody was raised against a synthetic peptide corresponding to 15 amino acids near the carboxy terminus of the avian influenza neuraminidase protein. Efforts were made to use relatively conserved regions as the antigen. The immunogen is located within the last 50 amino acids of Avian Influenza Neuraminidase. Amino Acid Squence: IRGRPKENTIWTSGS
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.