Learn More
Avian Influenza A H5N1 Hemagglutinin Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-41051MFV450
 View more versions of this product
Product Code. 18588637
  • £470.00 / 0.10mL
Description
Avian Influenza A H5N1 Hemagglutinin Polyclonal antibody specifically detects Avian Influenza A H5N1 Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Avian Influenza A H5N1 Hemagglutinin
Polyclonal
50 mM Sodium Borate
Rabbit
Peptide affinity purified
RUO
Primary
Store at 4°C in the dark.
IgG
Western Blot, ELISA
mFluor Violet 450 SE
Harvey rat sarcoma viral oncogene homolog
Antibody was raised against a synthetic peptide corresponding to 15 amino acids at the amino terminus of the avian influenza hemagglutinin protein. Efforts were made to use relatively conserved regions of the viral sequence as the antigen. The immunogen is located within amino acids 90 - 140 of Avian Influenza Hemagglutinin. Amino Acid Squence: CYPGDFNDYEELKHL
0.1 mL
Infections (Virus Bacteria and Parasites)
Virus
Purified