missing translation for 'onlineSavingsMsg'
Learn More

Avian Influenza A H5N1 Hemagglutinin Antibody - BSA Free, Novus Biologicals™

Goat Polyclonal Antibody

Brand:  Novus Biologicals NBP2-41065-0.025mg

 View more versions of this product

Product Code. 18699235

  • £150.00 / 0.03mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Avian Influenza Hemagglutinin 3 Polyclonal specifically detects Avian Influenza Hemagglutinin 3 in Virus samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Avian Influenza Hemagglutinin 3
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
Antibody was raised against a synthetic peptide corresponding to 14 amino acids near the center of the Hemagglutinin protein Efforts were made to use relatively conserved regions of the viral sequence as the antigen. The immunogen is located within amino acids 250 - 300 of Avian Influenza Hemagglutinin 3. Amino Acid Squence: APEYAYKIVKKGDS
0.025 mg
Primary
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Western Blot, ELISA
1 mg/mL
Western Blot, ELISA 1 μg/mL
Goat
Affinity Purified
RUO
Virus
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.