missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AVEN Rabbit anti-Human, Mouse, Rat, Clone: S06-7A9, Novus Biologicals™
Rabbit Monoclonal Antibody
£222.00 - £451.00
Specifications
| Antigen | AVEN |
|---|---|
| Clone | S06-7A9 |
| Dilution | Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunofluorescence 10 μg/mL, Immunocytochemistry 10 μg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18332536
|
Novus Biologicals
NBP3-15004-25UL |
25 μg |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18386875
|
Novus Biologicals
NBP3-15004-100UL |
100 μg |
£451.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AVEN Monoclonal antibody specifically detects AVEN in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, ImmunocytochemistrySpecifications
| AVEN | |
| Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunofluorescence 10 μg/mL, Immunocytochemistry 10 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| apoptosis, caspase activation inhibitor, cell death regulator Aven, PDCD12, programmed cell death 12 | |
| A synthetic peptide of human AVEN (Uniprot#: Q9NQS1) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S06-7A9 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| 57099 | |
| IgG | |
| Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title