Learn More
Invitrogen™ AVEN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588858
Description
Immunogen sequence: VQGTVPLEVP QVKPKRTDDG KGLGMQLKGP LGPGGRGPIF ELKSVAAGCP VLLGKDNPSP GPSRDSQKPT SPLQSAGDHL EEELDLLLNL DAPIKEGDNI LPDQTSQDLK SKEDGEVVQE EEVCAKPSVT EEKNMEPEQP STSKNVTEEE LEDWLDSMIS; Positive Samples: 22RV1, HT-29, LO2, HeLa; Cellular Location: Endomembrane system, Peripheral membrane protein.
Apoptosis plays a major role in normal organism development, tissue homeostasis, and removal of damaged cells. Disruption of this process has been implicated in a variety of diseases such as cancer. Aven is a recently discovered protein that blocks apoptosis induced by Apaf-1 and caspase-9. It is thought that Aven functions by binding to Bcl-xL, an antiapoptotic member of the Bcl-2 family, and to Apaf-1, possibly interfering with the ability of Apaf-1 to self-associate, suggesting that Aven impedes Apaf-1-mediated caspase activation. Higher levels of Aven mRNA are seen in patients with acute leukemia than in control patients, suggesting that Aven may be useful as a prognostic indicator in leukemia patients.
Specifications
| AVEN | |
| Polyclonal | |
| Unconjugated | |
| AVEN | |
| 1700013A01Rik; 1700056A21Rik; apoptosis and caspase activation inhibitor; apoptosis, caspase activation inhibitor; AVEN; Cell death regulator Aven; mAven-L; mAven-S; PDCD12; programmed cell death 12 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 311299, 57099 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.91 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9NQS1 | |
| AVEN | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 203-362 of human AVEN (NP_0651041). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.