missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AVEN Antibody (3G4), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-07098C
This item is not returnable.
View return policy
Description
AVEN Monoclonal antibody specifically detects AVEN in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| AVEN | |
| Monoclonal | |
| DyLight 650 | |
| NP_065104 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 3G4 | |
| 50mM Sodium Borate | |
| apoptosis, caspase activation inhibitor, cell death regulator Aven, PDCD12, programmed cell death 12 | |
| Recombinant human AVEN (254-362aa) purified from E. coli | |
| 0.1 mL | |
| Apoptosis | |
| 57099 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction