missing translation for 'onlineSavingsMsg'
Learn More

AVE3085, MedChemExpress

Product Code. 16491819
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491819 1 mg 1mg
16470248 5 mg 5mg
16427788 10 mg 10mg
16410606 20 mg 20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16491819 Supplier Medchem Express Supplier No. HY19504
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AVE3085 is a potent endothelial nitric oxide synthase enhancer, used for cardiovascular disease treatment.

AVE3085 is a potent endothelial nitric oxide synthase enhancer, used for cardiovascular disease treatment.

Specifications

Chemical Name or Material AVE3085
CAS 450348-85-3
Color White
Purity Grade Notes Research
Molecular Formula C17H13F2NO3
Quantity 1 mg
Solubility Information DMSO : 250 mg/mL (787.92 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(OC(F)(F)O2)C2=C1)NC3CC4=C(C=CC=C4)C3
Molecular Weight (g/mol) 317.29
Formula Weight 317.29
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.