missing translation for 'onlineSavingsMsg'
Learn More

Avasimibe, MedChemExpress

Product Code. 16471066
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471066 200 mg 200mg
16455558 10 mg 10mg
16446116 100 mg 100mg
16431727 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16471066 Supplier Medchem Express Supplier No. HY13215

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Avasimibe (CI-1011; PD-148515) is an orally active acyl coenzyme A-cholesterol acyltransferase (ACAT; also called SOAT)) inhibitor with IC50s of 24 and 9.2 μM for ACAT1 and ACAT2, respectively. Avasimibe can be used for the research of prostate cancer.

Avasimibe (CI-1011

Specifications

Chemical Name or Material Avasimibe
CAS 166518-60-1
Color White
Purity Grade Notes Research
Molecular Formula C29H43NO4S
Quantity 200 mg
Synonym CI-1011 PD-148515
Solubility Information DMSO : 250 mg/mL (498.29 mM; Need ultrasonic)
SMILES O=C(NS(=O)(OC1=C(C(C)C)C=CC=C1C(C)C)=O)CC2=C(C(C)C)C=C(C(C)C)C=C2C(C)C
Molecular Weight (g/mol) 501.72
Formula Weight 501.72
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.