missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Avagacestat
GREENER_CHOICE

Product Code. p-7161599
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18749576

Brand: Tocris Bioscience™ 6363/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly potent γ-secretase inhibitor; orally bioavailable

Avagacestat is a highly potent γ-secretase inhibitor (Aβ40 IC50 = 0.3 nM). Exhibits >137-fold selectivity for amyloid-β precursor protein (APP) processing over Notch. Reduces plasma and brain Aβ40 and Aβ42 levels in vivo. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2R)-2-[[(4-Chlorophenyl)sulfonyl][[2-fluoro-4-(1,2,4-oxadiazol-3-yl)phenyl]methyl]amino]-5,5,5-trifluoropentanamide
CAS 1146699-66-2
Synonym BMS-708163
Quantity 25 mg
Molecular Formula C20H17ClF4N4O4S
Formula Weight 520.89
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.