missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Avagacestat
Description
Avagacestat is a highly potent γ-secretase inhibitor (Aβ40 IC50 = 0.3 nM). Exhibits >137-fold selectivity for amyloid-β precursor protein (APP) processing over Notch. Reduces plasma and brain Aβ40 and Aβ42 levels in vivo. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | (2R)-2-[[(4-Chlorophenyl)sulfonyl][[2-fluoro-4-(1,2,4-oxadiazol-3-yl)phenyl]methyl]amino]-5,5,5-trifluoropentanamide |
| CAS | 1146699-66-2 |
| Synonym | BMS-708163 |
| Quantity | 5 mg |
| Molecular Formula | C20H17ClF4N4O4S |
| Formula Weight | 520.89 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?