missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Avagacestat

Product Code. 18749576
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18749576 25 mg 25mg
18787596 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18749576 Supplier Tocris Bioscience™ Supplier No. 6363/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly potent γ-secretase inhibitor; orally bioavailable

Avagacestat is a highly potent γ-secretase inhibitor (Aβ40 IC50 = 0.3 nM). Exhibits >137-fold selectivity for amyloid-β precursor protein (APP) processing over Notch. Reduces plasma and brain Aβ40 and Aβ42 levels in vivo. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2R)-2-[[(4-Chlorophenyl)sulfonyl][[2-fluoro-4-(1,2,4-oxadiazol-3-yl)phenyl]methyl]amino]-5,5,5-trifluoropentanamide
CAS 1146699-66-2
Synonym BMS-708163
Quantity 25 mg
Molecular Formula C20H17ClF4N4O4S
Formula Weight 520.89
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.