missing translation for 'onlineSavingsMsg'
Learn More

Avagacestat, MedChemExpress

Product Code. 16444027
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16444027

Brand: Medchem Express HY50845

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Avagacestat (BMS-708163) is a potent inhibitor of γ-secretase, with IC50s of 0.27 nM and 0.30 nM for Aβ42 and Aβ40 inhibition; Avagacestat (BMS-708163) also inhibits NICD (Notch IntraCellular Domain) with IC50 of 0.84 nM and shows weak inhibition of CYP2C19, with IC50 of 20 μM. Avagacestat can be used for Alzheimer disease research.

Avagacestat (BMS-708163) is a potent inhibitor of γ-secretase, with IC50s of 0.27 nM and 0.30 nM for Aβ42 and Aβ40 inhibition

Specifications

Chemical Name or Material Avagacestat
CAS 1146699-66-2
Color White
Purity Grade Notes Research
Molecular Formula C20H17ClF4N4O4S
Quantity 50 mg
Synonym BMS-708163
Solubility Information DMSO : ≥ 100 mg/mL (191.98 mM)
SMILES O=S(N([C@@H](C(N)=O)CCC(F)(F)F)CC1=CC=C(C2=NOC=N2)C=C1F)(C3=CC=C(Cl)C=C3)=O
Molecular Weight (g/mol) 520.89
Formula Weight 520.89
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.