missing translation for 'onlineSavingsMsg'
Learn More

AV-412, MedChemExpress

Product Code. 16473848
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16473848 10 mg 10mg
16426577 5 mg 5mg
16400227 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16473848 Supplier Medchem Express Supplier No. HY10346
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AV-412 (MP412) is an EGFR inhibitor with IC50s of 0.75, 0.5, 0.79, 2.3, 19 nM for EGFR, EGFRL858R, EGFRT790M, EGFRL858R/T790M and ErbB2, respectively.

AV-412 (MP412) is an EGFR inhibitor with IC50s of 0.75, 0.5, 0.79, 2.3, 19 nM for EGFR, EGFRL858R, EGFRT790M, EGFRL858R/T790M and ErbB2, respectively.

Specifications

Chemical Name or Material AV-412
CAS 451493-31-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C41H44ClFN6O7S2
Quantity 10 mg
Synonym MP412
Solubility Information DMSO : ≥ 28 mg/mL (32.89 mM)
SMILES O=S(C1=CC=C(C)C=C1)(O)=O.C=CC(NC2=CC3=C(NC4=CC=C(F)C(Cl)=C4)N=CN=C3C=C2C#CC(N5CCN(C)CC5)(C)C)=O.O=S(C6=CC=C(C)C=C6)(O)=O
Molecular Weight (g/mol) 851.41
Formula Weight 851.41
Percent Purity 98.1%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.