missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AUZ 454, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
2mg
50mg
5mg
Description
AUZ 454 (K03861) is a type II CDK2 inhibitor with Kd of 8.2 nM. AUZ 454 (K03861) inhibits CDK2 activity by competing with binding of activating cyclins.
Specifications
Specifications
| Chemical Name or Material | AUZ 454 |
| CAS | 853299-07-7 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C24H26F3N7O2 |
| Quantity | 5 mg |
| Synonym | K03861 |
| Solubility Information | DMSO : 250 mg/mL (498.50 mM; Need ultrasonic) |
| SMILES | NC1=NC(OC2=CC=C(NC(NC3=CC(C(F)(F)F)=C(CN4CCN(C)CC4)C=C3)=O)C=C2)=CC=N1 |
| Molecular Weight (g/mol) | 501.5 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction