missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AUTS2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP3-05291
This item is not returnable.
View return policy
Description
AUTS2 Polyclonal specifically detects AUTS2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA.
Specifications
| AUTS2 | |
| Polyclonal | |
| Flow Cytometry 10 ug/mL, Immunocytochemistry/Immunofluorescence 10 ug/mL, Peptide ELISA Detection limit 1:32000 | |
| autism susceptibility candidate 2, autism susceptibility gene 2 protein, autism-related protein 1, KIAA0442FBRSL2, MGC13140 | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunofluorescence, ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| 26053 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction