missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AUTS2 Antibody (CL7067), Novus Biologicals™
Mouse Monoclonal Antibody
£222.00 - £469.00
Specifications
| Antigen | AUTS2 |
|---|---|
| Clone | CL7067 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18647621
|
Novus Biologicals
NBP2-76529-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18362091
|
Novus Biologicals
NBP2-76529 |
100 μL |
£469.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AUTS2 Monoclonal specifically detects AUTS2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| AUTS2 | |
| Monoclonal | |
| Mouse | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| 26053 | |
| This antibody was developed against a synthetic peptide corresponding to amino acids: GISPLPGGERFPYPSFHWD | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| CL7067 | |
| Unconjugated | |
| Human, Mouse | |
| autism susceptibility candidate 2, autism susceptibility gene 2 protein, autism-related protein 1, KIAA0442FBRSL2, MGC13140 | |
| AUTS2 | |
| IgG1 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title