missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AUTS2 Antibody (CL7067) [HRP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76529H
This item is not returnable.
View return policy
Description
AUTS2 Monoclonal antibody specifically detects AUTS2 in Human,Mouse samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| AUTS2 | |
| Monoclonal | |
| HRP | |
| autism susceptibility candidate 2, autism susceptibility gene 2 protein, autism-related protein 1, KIAA0442FBRSL2, MGC13140 | |
| This antibody was developed using a synthetic peptide derived from Q8WXX7, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL7067 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 26053 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction