missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Autotaxin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14243-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene functions as both a phosphodiesterase, which cleaves phosphodiester bonds at the 5′ end of oligonucleotides, and a phospholipase, which catalyzes production of lysophosphatidic acid (LPA) in extracellular fluids. LPA evokes growth factor-like responses including stimulation of cell proliferation and chemotaxis. This gene product stimulates the motility of tumor cells and has angiogenic properties, and its expression is upregulated in several kinds of carcinomas. The gene product is secreted and further processed to make the biologically active form. Several alternatively spliced transcript variants encoding different isoforms have been identified.Specifications
| Autotaxin | |
| Polyclonal | |
| Unconjugated | |
| ENPP2 | |
| ATX, ATX X, Autotaxin, E NPP 2, ENPP2, LysoPLD, PD IALPHA, PDNP2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18606, 5168, 84050 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q13822, Q64610, Q9R1E6 | |
| Enpp2 | |
| Autotaxin Fusion Protein Ag5499 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction