Learn More
Cytiva AutoSeq G-50 Columns
Use for rapid removal of unincorporated fluorescent dye-terminators from cycle sequencing reactions prior to automated sequencing. Cytiva Life Sciences™ illustra™ AutoSeq G-50 Columns provide effective purification essential for high-quality sequencing results.
Brand:  Cytiva 27-5340-02
Code : GH
Additional Details : CAS Number : 9048-46-8 Weight : 0.87500kg
 View more versions of this product
Product Code. 11783309
  • £687.00 / Pack of 250
Estimated Shipment: 22-05-2024
to see stock.
Description
Description
  • Columns are prepacked with Sephadex G-50 DNA Grade F and pre-equilibrated in double-distilled water with 0.05% Kathon CG/ICP Biocide added as a preservative
  • Columns that are ready to use enable sample application to collection of purified product in less than 4 min.
  • Can be used to clean up reactions prepared with the BigDye Terminator v3.1 Cycle Sequencing Kit from Applied Biosystems
  • Typical sample volume 12 to 25 μl
  • Suitable for purification of DNA greater than 20 bases in length
  • Optimized for automated fluorescent sequencing
Specifications
Specifications
Rapid removal of unincorporated Fluorescent dye-terminators from cycle sequencing reactions prior to automated sequencing
Column
250 Columns