Learn More
Hamilton™ Autosampler Syringes
Product Code. 10156441
Change view
Click to view available options
For Use With (Equipment):
AS200 Autosampler
Thermo-Finnigan / Fisons AS800/2000
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10156441 AS200 Autosampler Each
11537714 Thermo-Finnigan / Fisons AS800/2000 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10156441 Supplier Hamilton™ Supplier No. 202201
Only null left
Add to basket
Add to basket
Specifications
Material Stainless Steel (Plunger)
Needle Gauge 25s G
Diameter (Metric) Inner 0.48 mm
Diameter (Metric) Outer 6.6 mm
Product Type Autosampler Syringe
Model 701
Needle Point Number 2
Quantity 1/Ea.
Volume (Metric) 10 μL
Needle Point Style 2
For Use With (Equipment) AS200 Autosampler
Connection Type Fixed