Learn More
Trajan Scientific and Medical™ Autosampler Syringes with Fixed Needle for Thermo Scientific GCs
Gives better performance and superior quality. Trajan Scientific and Medical Autosampler Syringes with Fixed Needle for Thermo Scientific GCs has the knowledge and expertise to manufacture an SGE syringe.
Brand: Trajan Scientific and Medical™ 002869
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Specifically designed for Thermo Scientific instruments
Specifications
CTC RTC and Thermo RSH Syringe | |
0.47 mm | |
10 μL | |
Thermo Gas Chromatography |
26 G | |
Fixed Needle | |
Bevel |