missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ Autosampler Syringes for Agilent GCs
Gives better performance and superior quality. Trajan Scientific and Medical Autosampler Syringes for Agilent GCs has the knowledge and expertise to manufacture an SGE syringe.
£55.30 - £1370.00
Product Code | Brand | Product Type | Volume (Metric) | Needle Gauge | Diameter (Metric) Outer Needle | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Volume (Metric) | Needle Gauge | Diameter (Metric) Outer Needle | Price | Quantity & Availability | ||
15258453
|
Trajan Scientific and Medical™
002816 |
Fixed Needle | 10 μL | 23 G | 0.63 mm |
£1370.00
Pack of 25 |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10390371
|
Trajan Scientific and Medical™
001800 |
Fixed Needle | 5 μL | 26 G | 0.47 mm |
£71.70
Each |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10063622
|
Trajan Scientific and Medical™
002826 |
Fixed Needle with GT Plunger | 10 μL | 23 and 26 G | 0.63, 0.47 mm |
£65.30
Each |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
15248453
|
Trajan Scientific and Medical™
002811 |
Fixed Needle | 10 μL | 23 G | 0.63 mm |
£55.30
Each |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Specifically designed for Agilent instruments
Specifications
Agilent Syringe |