Learn More
Merck Autorad Orientation Marker
Product Code. 10173783 Shop All Merck Products
Change view
Click to view available options
Quantity:
5 Pk.
Unit Size:
Pack of 5
Product Code. Quantity unitSize
10173783 5 Pk. Pack of 5
1 options
Product Code. 10173783 Supplier Merck Supplier No. 20135
Autorad markers ensure the accurate orientation of an autoradiograph to its membrane, emitting low-level luminescence rather than radioactivity making them a safe, non-hazardous alternative to radioactive ink.
  • For accurate orientation of an autoradiograph to its membrane
  • Emits low-level luminescence rather than radioactivity
  • Safe, non-hazardous alternative to radioactive ink
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Used in Western Blotting, dot/slot Blotting, For Use on Immunoblots
Quantity 5 Pk.
Product Type Orientation Marker