Learn More
Biochrom Automatic Sipper System
Automatic Sipper System for Ultrospec 7000/8000/9000. Aspiration system using a peristaltic pump head designed to automatically read a sample from a tube, beaker or flask.
Brand: Biochrom 80-7100-36
Code : GE
Additional Details : Weight : 2.00000kg
Description
- Automatic aspiration
- User defined air gap and sip volume
- Waste or return option
- Local or PC-controlled
- Ability to calibrate sipped volume
- Choice of tubing kits
- Requires Tubing kit 80-7100-37 or 80-7100-38 and suitable flowcell 80-7100-40
Specifications
Automatic Sipper System | |
Ultrospec 7000/8000/9000 |